siRNA Details
| Gene Symbol: CCNYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142300.1 | NP_001135772.1 | 3 | 673 | ||
| NM_152523.2 | NP_689736.1 | 4 | 421 | |||
| XM_005246343.1 | XP_005246400.1 | 3 | 439 | |||
| GenBank mRNA | AK097751.1 | - | 4 | 421 | ||
| BC067253.1 | - | 3 | 460 | 6500304 | ||
| CN482952.1 | - | 3 | 561 | |||

CCNYL1
Genomic Map