siRNA Details
| Gene Symbol: FGD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320276.1 | NP_001307205.1 | 1 | 2190 | ||
| NM_152536.3 | NP_689749.3 | 1 | 2190 | |||
| XM_011533422.1 | XP_011531724.1 | 1 | 2190 | |||
| XM_011533423.1 | XP_011531725.1 | 1 | 2190 | |||
| GenBank mRNA | AK097276.1 | - | 1 | 703 | ||
| AK131078.1 | - | 1 | 1854 | |||
| BC132814.1 | - | 1 | 1828 | 40146836 | ||
| BC132816.1 | - | 1 | 1828 | 40146838 | ||
| BC144300.1 | - | 1 | 1828 | 9052820 | ||
| BX538312.1 | - | 1 | 331 | |||
| BX640820.1 | - | 1 | 1131 | |||

FGD5
Genomic Map