siRNA Details
| Gene Symbol: SLC35G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152462.2 | NP_689675.1 | 1 | 1185 | ||
| GenBank mRNA | AK097473.1 | - | 1 | 1150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC35G3| Gene Symbol: SLC35G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152462.2 | NP_689675.1 | 1 | 1185 | ||
| GenBank mRNA | AK097473.1 | - | 1 | 1150 | ||