siRNA Details
| Gene Symbol: C19orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152482.2 | NP_689695.2 | 3 | 676 | ||
| XM_005259506.3 | XP_005259563.1 | 3 | 692 | |||
| XM_006722653.3 | XP_006722716.1 | 3 | 651 | |||
| XM_017026374.1 | XP_016881863.1 | 3 | 999 | |||
| GenBank mRNA | AK056004.1 | - | 1 | 1704 | ||
| AK075267.1 | - | 3 | 659 | |||
| AK093985.1 | - | 4 | 553 | |||
| AK297944.1 | - | 3 | 784 | |||
| AL122089.1 | - | 3 | 647 | |||
| AY007120.1 | - | 2 | 612 | |||
| BC018441.1 | - | 3 | 646 | 4471390 | ||

C19orf25
Genomic Map