siRNA Details
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114938.2 | NP_001108410.2 | 4 | 722 | ||
NM_001190182.1 | NP_001177111.1 | 4 | 695 | |||
XM_011540812.1 | XP_011539114.1 | 3 | 1549 | |||
XM_011540814.1 | XP_011539116.1 | 3 | 1549 | |||
XM_011540820.1 | XP_011539122.1 | 3 | 1549 | |||
XM_017000450.1 | XP_016855939.1 | 3 | 1522 | |||
XM_017000451.1 | XP_016855940.1 | 2 | 1453 | |||
XM_017000452.1 | XP_016855941.1 | 4 | 1450 | |||
XM_017000453.1 | XP_016855942.1 | 3 | 1549 | |||
XM_017000454.1 | XP_016855943.1 | 3 | 620 | |||
XM_017000455.1 | XP_016855944.1 | 2 | 167 | |||
XM_017000456.1 | XP_016855945.1 | 2 | 168 | |||
XM_017000457.1 | XP_016855946.1 | 3 | 1549 | |||
XM_017000458.1 | XP_016855947.1 | 3 | 1549 | |||
GenBank mRNA | AK057646.1 | - | 3 | 625 | ||
AK298104.1 | - | 4 | 693 | |||
AK303939.1 | - | 4 | 720 | |||
AK310879.1 | - | 2 | 928 | |||
BC029888.1 | - | 2 | 345 | 5175485 | ||
BC128177.1 | - | 2 | 311 | 40113154 |
