siRNA Details
Gene Symbol: LINC00479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027272.1 | 5 | 530 | |||
GenBank mRNA | AK057397.1 | - | 5 | 530 | ||
BC128198.1 | - | 5 | 551 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027272.1 | 5 | 530 | |||
GenBank mRNA | AK057397.1 | - | 5 | 530 | ||
BC128198.1 | - | 5 | 551 |