siRNA Details
| Gene Symbol: XXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152531.4 | NP_689744.3 | 1, 2 | 620 | ||
| XM_011512448.2 | XP_011510750.1 | 2, 3 | 432 | |||
| XM_017005749.1 | XP_016861238.1 | 1, 2 | 613 | |||
| GenBank mRNA | AK075551.1 | - | 1, 2 | 588 | ||
| BC019036.2 | - | 1, 2 | 529 | 4591786 | ||
| BC039067.1 | - | 1, 2 | 602 | 5185557 | ||

XXYLT1
Genomic Map