siRNA Details
Gene Symbol: SLC35G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152462.2 | NP_689675.1 | 1 | 1590 | ||
GenBank mRNA | AK097473.1 | - | 1 | 1555 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC35G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152462.2 | NP_689675.1 | 1 | 1590 | ||
GenBank mRNA | AK097473.1 | - | 1 | 1555 |