siRNA Details
| Gene Symbol: ZNF565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042474.2 | NP_001035939.1 | 5 | 1787 | ||
| NM_152477.4 | NP_689690.3 | 5 | 1642 | |||
| XM_011526512.2 | XP_011524814.1 | 5 | 1855 | |||
| XM_011526514.2 | XP_011524816.1 | 5 | 1721 | |||
| XM_017026341.1 | XP_016881830.1 | 6 | 2330 | |||
| XM_017026342.1 | XP_016881831.1 | 4 | 1550 | |||
| XM_017026343.1 | XP_016881832.1 | 4 | 1596 | |||
| GenBank mRNA | AK057290.1 | - | 5 | 2374 | ||
| AK094310.1 | - | 5 | 1784 | |||
| BC068453.1 | - | 5 | 1624 | 30343899 | ||

ZNF565
Genomic Map