siRNA Details
| Gene Symbol: XXYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308069.1 | NP_001294998.1 | 4 | 1283 | ||
| NM_152531.4 | NP_689744.3 | 4 | 1746 | |||
| XM_005269286.4 | XP_005269343.1 | 4 | 4131 | |||
| XM_011512448.2 | XP_011510750.1 | 5 | 1558 | |||
| XM_017005750.1 | XP_016861239.1 | 3 | 4519 | |||
| GenBank mRNA | AK057046.1 | - | 5 | 1491 | ||
| AK075551.1 | - | 4 | 1714 | |||
| AK092474.1 | - | 2 | 1023 | |||
| BC019036.2 | - | 4 | 1655 | 4591786 | ||
| BC039067.1 | - | 4 | 1728 | 5185557 | ||

XXYLT1
Genomic Map