siRNA Details
| Gene Symbol: CCDC112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040440.2 | NP_001035530.1 | 3, 4 | 576 | ||
| NM_152549.2 | NP_689762.2 | 3, 4 | 622 | |||
| XM_005271908.1 | XP_005271965.1 | 3, 4 | 570 | |||
| GenBank mRNA | AJ580807.1 | - | 3, 4 | 475 | ||
| BC031242.1 | - | 3, 4 | 600 | 5265792 | ||

CCDC112
Genomic Map