siRNA Details
| Gene Symbol: SYNE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152592.3 | NP_689805.3 | 12 | 2245 | ||
| XM_005267377.3 | XP_005267434.1 | 13 | 2342 | |||
| XM_006720063.3 | XP_006720126.1 | 13 | 2442 | |||
| XM_011536515.2 | XP_011534817.1 | 13 | 2377 | |||
| XM_017021056.1 | XP_016876545.1 | 12 | 2244 | |||
| GenBank mRNA | AK098471.1 | - | 7 | 1008 | ||
| AK131436.1 | - | 13 | 2110 | |||
| BC146604.1 | - | 13 | 2245 | 40123293 | ||
| BX648302.1 | - | 10 | 4117 | |||

SYNE3
Genomic Map