siRNA Details
Gene Symbol: ZNF579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152600.2 | NP_689813.2 | 2 | 1696 | ||
GenBank mRNA | AK092772.1 | - | 2 | 1672 | ||
BC042926.1 | - | 1 | 534 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152600.2 | NP_689813.2 | 2 | 1696 | ||
GenBank mRNA | AK092772.1 | - | 2 | 1672 | ||
BC042926.1 | - | 1 | 534 |