siRNA Details
| Gene Symbol: C2orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152614.2 | NP_689827.2 | 1 | 237 | ||
| GenBank mRNA | BC034405.1 | - | 1 | 200 | 5169625 | |
| BC063389.1 | - | 1 | 200 | 5170341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf57| Gene Symbol: C2orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152614.2 | NP_689827.2 | 1 | 237 | ||
| GenBank mRNA | BC034405.1 | - | 1 | 200 | 5169625 | |
| BC063389.1 | - | 1 | 200 | 5170341 | ||