siRNA Details
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286441.1 | NP_001273370.1 | 6 | 736 | ||
| NM_152596.3 | NP_689809.2 | 4 | 481 | |||
| XM_006720415.3 | XP_006720478.1 | 5 | 653 | |||
| XM_011521298.2 | XP_011519600.1 | 7 | 966 | |||
| XM_011521299.2 | XP_011519601.1 | 7 | 697 | |||
| XM_011521300.2 | XP_011519602.1 | 4 | 225 | |||
| XM_011521301.2 | XP_011519603.1 | 6 | 522 | |||
| XM_017021970.1 | XP_016877459.1 | 4 | 225 | |||
| XM_017021971.1 | XP_016877460.1 | 4 | 223 | |||
| GenBank mRNA | AK292524.1 | - | 4 | 387 | ||
| AK302864.1 | - | 6 | 670 | |||
| BC030628.2 | - | 4 | 421 | 4827307 | ||

EXD1
Genomic Map