siRNA Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324338.1 | NP_001311267.1 | 2, 3 | 244 | ||
| NM_152597.4 | NP_689810.3 | 2, 3 | 341 | |||
| XM_011521305.2 | XP_011519607.1 | 2, 3 | 217 | |||
| XM_011521306.1 | XP_011519608.1 | 2, 3 | 630 | |||
| XM_011521307.2 | XP_011519609.1 | 2, 3 | 217 | |||
| XM_011521309.2 | XP_011519611.1 | 2, 3 | 217 | |||
| GenBank mRNA | AK093308.1 | - | 2, 3 | 323 | ||
| AY260140.1 | - | 1, 2 | 138 | |||
| BC045191.2 | - | 2, 3 | 333 | 5297527 | ||
| BX642685.1 | - | 2, 3 | 342 | |||

FSIP1
Genomic Map