siRNA Details
| Gene Symbol: CCER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152638.3 | NP_689851.1 | 1 | 1183 | ||
| GenBank mRNA | BC024183.2 | - | 1 | 1181 | 4822259 | |
| BC026075.2 | - | 1 | 1181 | 4828455 | ||
| BC027600.2 | - | 1 | 1183 | 4828721 | ||
| BC034491.2 | - | 1 | 1181 | 4822257 | ||

CCER1
Genomic Map