siRNA Details
| Gene Symbol: FAM227B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152647.2 | NP_689860.2 | 7 | 759 | ||
| XM_005254213.3 | XP_005254270.1 | 7 | 836 | |||
| XM_005254214.3 | XP_005254271.1 | 7 | 769 | |||
| XM_005254215.3 | XP_005254272.1 | 6 | 800 | |||
| XM_005254216.3 | XP_005254273.1 | 7 | 836 | |||
| XM_005254219.3 | XP_005254276.1 | 7 | 836 | |||
| XM_005254220.3 | XP_005254277.1 | 7 | 836 | |||
| XM_006720423.2 | XP_006720486.1 | 8 | 801 | |||
| XM_006720424.2 | XP_006720487.1 | 7 | 836 | |||
| XM_006720427.3 | XP_006720490.1 | 8 | 807 | |||
| XM_011521319.2 | XP_011519621.1 | 7 | 2719 | |||
| XM_011521320.1 | XP_011519622.1 | 5 | 211 | |||
| XM_011521321.2 | XP_011519623.1 | 7 | 836 | |||
| XM_011521322.1 | XP_011519624.1 | 4 | 175 | |||
| XM_011521324.1 | XP_011519626.1 | 7 | 836 | |||
| XM_011521325.2 | XP_011519627.1 | 7 | 836 | |||
| XM_011521326.1 | XP_011519628.1 | 7 | 836 | |||
| XM_017021991.1 | XP_016877480.1 | 8 | 876 | |||
| XM_017021992.1 | XP_016877481.1 | 5 | 524 | |||
| XM_017021995.1 | XP_016877484.1 | 7 | 836 | |||
| XM_017021996.1 | XP_016877485.1 | 6 | 284 | |||
| GenBank mRNA | AK057362.1 | - | 7 | 757 | ||
| BC048125.1 | - | 7 | 761 | 5272014 | ||
| BC048139.1 | - | 7 | 761 | |||

FAM227B
Genomic Map