siRNA Details
| Gene Symbol: LINC01600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131168.1 | 3 | 2322 | |||
| GenBank mRNA | AK056496.1 | - | 3 | 2321 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01600| Gene Symbol: LINC01600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131168.1 | 3 | 2322 | |||
| GenBank mRNA | AK056496.1 | - | 3 | 2321 | ||