siRNA Details
| Gene Symbol: C11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152587.3 | NP_689800.3 | 8, 9 | 865 | ||
| XM_005271412.3 | XP_005271469.1 | 7, 8 | 827 | |||
| XM_005271413.3 | XP_005271470.1 | 7, 8 | 788 | |||
| XM_011542639.2 | XP_011540941.1 | 9, 10 | 918 | |||
| XM_011542641.2 | XP_011540943.1 | 6, 7 | 718 | |||
| XM_017017246.1 | XP_016872735.1 | 8, 9 | 825 | |||
| XM_017017247.1 | XP_016872736.1 | 8, 9 | 881 | |||
| GenBank mRNA | BC029536.1 | - | 8, 9 | 838 | 5298158 | |

C11orf65
Genomic Map