siRNA Details
| Gene Symbol: LMNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145727.2 | NP_001139199.1 | 4 | 502 | ||
| NM_001145728.2 | NP_001139200.1 | 6 | 988 | |||
| NM_001256266.1 | NP_001243195.1 | 6 | 900 | |||
| NM_152590.3 | NP_689803.2 | 5 | 869 | |||
| XM_011520575.1 | XP_011518877.1 | 8 | 1094 | |||
| XM_011520576.2 | XP_011518878.1 | 8 | 1094 | |||
| XM_011520577.2 | XP_011518879.1 | 8 | 1504 | |||
| XM_011520578.2 | XP_011518880.1 | 8 | 1117 | |||
| XM_011520579.2 | XP_011518881.1 | 8 | 1094 | |||
| XM_011520582.1 | XP_011518884.1 | 8 | 1106 | |||
| XM_011520583.1 | XP_011518885.1 | 7 | 975 | |||
| XM_011520584.1 | XP_011518886.1 | 5 | 782 | |||
| XM_011520585.2 | XP_011518887.1 | 5 | 705 | |||
| XM_011520587.1 | XP_011518889.1 | 5 | 478 | |||
| XM_011520588.1 | XP_011518890.1 | 4 | 400 | |||
| XM_017018891.1 | XP_016874380.1 | 7 | 926 | |||
| XM_017018892.1 | XP_016874381.1 | 7 | 996 | |||
| XM_017018893.1 | XP_016874382.1 | 6 | 805 | |||
| XM_017018894.1 | XP_016874383.1 | 8 | 1094 | |||
| GenBank mRNA | AK093323.1 | - | 5 | 830 | ||
| AK296816.1 | - | 4 | 494 | |||
| AK302292.1 | - | 6 | 988 | |||
| BC037957.1 | - | 6 | 869 | 5744078 | ||

LMNTD1
Genomic Map