siRNA Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324338.1 | NP_001311267.1 | 5 | 630 | ||
| NM_152597.4 | NP_689810.3 | 5 | 727 | |||
| XM_011521305.2 | XP_011519607.1 | 5 | 603 | |||
| XM_011521306.1 | XP_011519608.1 | 5 | 1016 | |||
| XM_011521307.2 | XP_011519609.1 | 5 | 603 | |||
| XM_011521309.2 | XP_011519611.1 | 5 | 603 | |||
| GenBank mRNA | AK093308.1 | - | 5 | 709 | ||
| AY260140.1 | - | 4 | 524 | |||
| BC045191.2 | - | 5 | 719 | 5297527 | ||

FSIP1
Genomic Map