siRNA Details
| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152613.2 | NP_689826.2 | 6 | 2301 | ||
| GenBank mRNA | BC022546.1 | - | 6 | 2238 | 4811804 | |
| BC022549.1 | - | 6 | 2237 | 4815849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WBP2NL| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152613.2 | NP_689826.2 | 6 | 2301 | ||
| GenBank mRNA | BC022546.1 | - | 6 | 2238 | 4811804 | |
| BC022549.1 | - | 6 | 2237 | 4815849 | ||