siRNA Details
| Gene Symbol: FAM46D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170574.1 | NP_001164045.1 | 5 | 1374 | ||
| NM_152630.4 | NP_689843.1 | 3 | 1298 | |||
| GenBank mRNA | AK128147.1 | - | 5 | 1374 | ||
| AK313875.1 | - | 3 | 1269 | |||
| BC028710.2 | - | 3 | 1267 | 4827590 | ||
| BC030784.2 | - | 3 | 1279 | 4826421 | ||
| BC034979.2 | - | 3 | 1264 | 4824369 | ||
| BX537938.1 | - | 3 | 1300 | |||

FAM46D
Genomic Map