siRNA Details
| Gene Symbol: FANCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018113.2 | NP_001018123.1 | 3 | 844 | ||
| NM_001324162.1 | NP_001311091.1 | 3 | 823 | |||
| NM_152633.3 | NP_689846.1 | 2 | 723 | |||
| XM_011545470.2 | XP_011543772.1 | 3 | 1030 | |||
| XM_017029355.1 | XP_016884844.1 | 3 | 922 | |||
| XM_017029356.1 | XP_016884845.1 | 3 | 1030 | |||
| GenBank mRNA | AK091383.1 | - | 2 | 723 | ||
| BC043596.1 | - | 3 | 827 | |||
| BC136558.1 | - | 3 | 826 | 9020548 | ||
| BC136560.1 | - | 2 | 705 | 9020550 | ||

FANCB
Genomic Map