siRNA Details
| Gene Symbol: PLD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195811.1 | NP_001182740.1 | 3 | 303 | ||
| NM_001320272.1 | NP_001307201.1 | 4 | 597 | |||
| NM_152666.2 | NP_689879.2 | 4 | 590 | |||
| XM_006711752.2 | XP_006711815.1 | 3 | 757 | |||
| XM_011544115.2 | XP_011542417.1 | 5 | 665 | |||
| XM_011544116.2 | XP_011542418.1 | 5 | 493 | |||
| XM_011544120.2 | XP_011542422.1 | 3 | 334 | |||
| XM_017000567.1 | XP_016856056.1 | 3 | 770 | |||
| XM_017000568.1 | XP_016856057.1 | 3 | 291 | |||
| XM_017000569.1 | XP_016856058.1 | 3 | 2295 | |||
| GenBank mRNA | AK098092.1 | - | 4 | 597 | ||
| AK295421.1 | - | 3 | 284 | |||
| AK307857.1 | - | 3 | 282 | |||
| AY391836.1 | - | 2 | 153 | |||
| AY461578.1 | - | 3 | 303 | |||
| BC101373.1 | - | 3 | 230 | 40025860 | ||
| BC101374.1 | - | 3 | 230 | 40025861 | ||
| BC101375.1 | - | 3 | 230 | 40025862 | ||
| DR004966.1 | - | 3 | 605 | |||

PLD5
Genomic Map