siRNA Details
| Gene Symbol: RWDD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307922.1 | NP_001294851.1 | 4 | 526 | ||
| NM_152682.3 | NP_689895.2 | 4 | 530 | |||
| GenBank mRNA | AK021702.1 | - | 4 | 454 | ||
| AK293274.1 | - | 2 | 271 | |||
| AK315515.1 | - | 4 | 464 | |||
| AL540937.3 | - | 4 | 467 | |||
| AL559547.3 | - | 4 | 463 | |||
| BC017472.1 | - | 4 | 465 | 3909581 | ||
| BC067752.1 | - | 4 | 489 | 5270461 | ||
| BC107432.1 | - | 4 | 311 | 6104330 | ||
| DA766837.1 | - | 4 | 529 | |||

RWDD4
Genomic Map