siRNA Details
| Gene Symbol: TAPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093550.1 | - | 1 | 727 | ||
| AK096972.1 | - | 1 | 1104 | |||
| BC132927.1 | - | 1 | 1104 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAPT1-AS1| Gene Symbol: TAPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093550.1 | - | 1 | 727 | ||
| AK096972.1 | - | 1 | 1104 | |||
| BC132927.1 | - | 1 | 1104 | |||