siRNA Details
| Gene Symbol: DOK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152721.5 | NP_689934.2 | 5 | 724 | ||
| XM_011525875.2 | XP_011524177.1 | 5 | 745 | |||
| XM_017025610.1 | XP_016881099.1 | 3 | 2785 | |||
| XM_017025611.1 | XP_016881100.1 | 3 | 1299 | |||
| GenBank mRNA | AK057795.1 | - | 4 | 433 | ||
| AY599248.1 | - | 5 | 722 | |||
| BC008583.1 | - | 3 | 306 | |||
| BC019045.2 | - | 3 | 349 | |||
| BC096744.1 | - | 5 | 717 | 5260167 | ||

DOK6
Genomic Map