siRNA Details
| Gene Symbol: CPNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152727.5 | NP_689940.3 | 3 | 549 | ||
| GenBank mRNA | AF492484.1 | - | 3 | 500 | ||
| AK292269.1 | - | 2 | 458 | |||
| AK315394.1 | - | 3 | 452 | |||
| BC053658.1 | - | 4 | 623 | 5750192 | ||
| BG830959.1 | - | 3 | 495 | |||
| BG831872.1 | - | 3 | 497 | |||
| CR749617.1 | - | 2 | 210 | |||

CPNE2
Genomic Map