siRNA Details
| Gene Symbol: BTBD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099272.1 | NP_001092742.1 | 3 | 708 | ||
| NM_001172418.1 | NP_001165889.1 | 2 | 493 | |||
| NM_052893.1 | NP_443125.1 | 4 | 908 | |||
| NM_152733.2 | NP_689946.2 | 2 | 445 | |||
| XM_011514279.2 | XP_011512581.1 | 5 | 1305 | |||
| XM_011514280.2 | XP_011512582.1 | 3 | 986 | |||
| XM_011514281.2 | XP_011512583.1 | 3 | 986 | |||
| XM_011514282.2 | XP_011512584.1 | 3 | 986 | |||
| XM_017010227.1 | XP_016865716.1 | 4 | 1186 | |||
| GenBank mRNA | AB067467.1 | - | 3 | 574 | ||
| AK057507.1 | - | 2 | 445 | |||
| BC101354.2 | - | 2 | 366 | 40025568 | ||
| BC101355.1 | - | 2 | 366 | 40025569 | ||
| BC101356.2 | - | 2 | 366 | |||
| BC101357.1 | - | 2 | 414 | 40025572 | ||
| BX537534.1 | - | 3 | 441 | |||

BTBD9
Genomic Map