siRNA Details
| Gene Symbol: SDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079653.1 | NP_001073121.1 | 15 | 2203 | ||
| NM_152744.3 | NP_689957.3 | 41 | 6058 | |||
| XM_011515188.1 | XP_011513490.1 | 35 | 4899 | |||
| XM_011515189.1 | XP_011513491.1 | 35 | 4896 | |||
| XM_017011837.1 | XP_016867326.1 | 35 | 4899 | |||
| GenBank mRNA | AK055987.1 | - | 2 | 129 | ||
| AK074077.1 | - | 8 | 998 | |||
| AK074083.1 | - | 26 | 3694 | |||
| AL833354.1 | - | 15 | 2200 | |||
| AY310398.1 | - | 41 | 6058 | |||

SDK1
Genomic Map