siRNA Details
| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 1, 2 | 470 | ||
| GenBank mRNA | AK096336.1 | - | 1, 2 | 164 | ||
| AK294261.1 | - | 1, 2 | 120 | |||
| AK308800.1 | - | 1, 2 | 176 | |||
| AK315486.1 | - | 1, 2 | 213 | |||
| BC028400.2 | - | 1, 2 | 194 | 4816822 | ||
| DC344091.1 | - | 1, 2 | 471 | |||

C8orf46
Genomic Map