siRNA Details
| Gene Symbol: TCTEX1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152665.2 | NP_689878.2 | 4, 5 | 475 | ||
| XM_011540933.2 | XP_011539235.1 | 5, 6 | 862 | |||
| XM_017000566.1 | XP_016856055.1 | 4, 5 | 771 | |||
| GenBank mRNA | AK098192.1 | - | 4, 5 | 473 | ||
| AK307100.1 | - | 6, 7 | 567 | |||
| BC106885.2 | - | 4, 5 | 440 | 40029630 | ||
| DA520308.1 | - | 4, 5 | 475 | |||

TCTEX1D1
Genomic Map