siRNA Details
| Gene Symbol: ZNF449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833407.1 | - | 2, 3 | 503 | ||
| AY280801.1 | - | 2, 3 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF449| Gene Symbol: ZNF449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833407.1 | - | 2, 3 | 503 | ||
| AY280801.1 | - | 2, 3 | 503 | |||