siRNA Details
| Gene Symbol: FBXO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142958.1 | NP_001136430.1 | 6, 7 | 1003 | ||
| NM_152676.2 | NP_689889.1 | 6, 7 | 1031 | |||
| XM_011525856.1 | XP_011524158.1 | 5, 6 | 762 | |||
| XM_011525859.1 | XP_011524161.1 | 5, 6 | 845 | |||
| XM_017025603.1 | XP_016881092.1 | 7, 8 | 1013 | |||
| XM_017025604.1 | XP_016881093.1 | 6, 7 | 3041 | |||
| GenBank mRNA | AK094215.1 | - | 6, 7 | 943 | ||
| BC029579.1 | - | 6, 7 | 981 | 5268841 | ||

FBXO15
Genomic Map