siRNA Details
Gene Symbol: TMEM192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100389.1 | NP_001093859.1 | 3 | 357 | ||
XM_011531717.2 | XP_011530019.1 | 3 | 398 | |||
XM_011531718.2 | XP_011530020.1 | 3 | 367 | |||
GenBank mRNA | AK095801.1 | - | 4 | 429 | ||
BC036301.1 | - | 3 | 302 | 4778254 | ||
BX538029.1 | - | 3 | 315 | |||
BX648915.1 | - | 3 | 313 |
