siRNA Details
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093550.1 | - | 1 | 857 | ||
AK096972.1 | - | 1 | 1234 | |||
BC132927.1 | - | 1 | 1234 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093550.1 | - | 1 | 857 | ||
AK096972.1 | - | 1 | 1234 | |||
BC132927.1 | - | 1 | 1234 |