siRNA Details
| Gene Symbol: AMER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105193.1 | NP_001098663.1 | 2 | 1337 | ||
| NM_001105194.1 | NP_001098664.1 | 2 | 1273 | |||
| NM_001105195.1 | NP_001098665.1 | 2 | 1270 | |||
| NM_152698.2 | NP_689911.2 | 2 | 1353 | |||
| XM_011510793.2 | XP_011509095.1 | 2 | 2379 | |||
| GenBank mRNA | AK095696.1 | - | 2 | 1317 | ||
| AK289525.1 | - | 2 | 1422 | |||
| BC113445.1 | - | 2 | 1230 | 8322497 | ||
| BC113447.1 | - | 2 | 1230 | 8322499 | ||
| BC143806.1 | - | 2 | 1227 | 9052321 | ||

AMER3
Genomic Map