siRNA Details
| Gene Symbol: CPNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152727.5 | NP_689940.3 | 9 | 1106 | ||
| GenBank mRNA | AF492484.1 | - | 9 | 1057 | ||
| AK292269.1 | - | 8 | 1015 | |||
| AK315394.1 | - | 9 | 1009 | |||
| BC053658.1 | - | 10 | 1180 | 5750192 | ||
| CR749617.1 | - | 8 | 767 | |||

CPNE2
Genomic Map