siRNA Details
| Gene Symbol: BTBD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099272.1 | NP_001092742.1 | 11 | 2076 | ||
| NM_001172418.1 | NP_001165889.1 | 11 | 1948 | |||
| NM_052893.1 | NP_443125.1 | 12 | 2276 | |||
| NM_152733.2 | NP_689946.2 | 10 | 1813 | |||
| XM_011514279.2 | XP_011512581.1 | 13 | 2673 | |||
| GenBank mRNA | AB067467.1 | - | 11 | 1942 | ||
| AK057507.1 | - | 10 | 1813 | |||
| AK090930.1 | - | 7 | 780 | |||
| BC101354.2 | - | 10 | 1734 | 40025568 | ||
| BC101355.1 | - | 10 | 1734 | 40025569 | ||
| BC101356.2 | - | 11 | 1803 | |||
| BC101357.1 | - | 11 | 1869 | 40025572 | ||
| BX537534.1 | - | 11 | 1807 | |||

BTBD9
Genomic Map