siRNA Details
| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 6 | 2659 | ||
| GenBank mRNA | AK055047.1 | - | 3 | 1993 | ||
| AK096336.1 | - | 7 | 2447 | |||
| BC028400.2 | - | 6 | 2384 | 4816822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8orf46| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 6 | 2659 | ||
| GenBank mRNA | AK055047.1 | - | 3 | 1993 | ||
| AK096336.1 | - | 7 | 2447 | |||
| BC028400.2 | - | 6 | 2384 | 4816822 | ||