siRNA Details
| Gene Symbol: CATSPERD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152784.3 | NP_689997.3 | 16 | 1545 | ||
| XM_011527885.2 | XP_011526187.1 | 16 | 1837 | |||
| XM_011527886.2 | XP_011526188.1 | 16 | 1783 | |||
| XM_011527887.2 | XP_011526189.1 | 16 | 1837 | |||
| XM_011527888.2 | XP_011526190.1 | 16 | 1837 | |||
| XM_011527889.2 | XP_011526191.1 | 16 | 1837 | |||
| XM_011527890.2 | XP_011526192.1 | 16 | 1837 | |||
| XM_011527891.1 | XP_011526193.1 | 7 | 633 | |||
| XM_011527892.1 | XP_011526194.1 | 5 | 354 | |||
| XM_017026564.1 | XP_016882053.1 | 15 | 1753 | |||
| XM_017026565.1 | XP_016882054.1 | 15 | 1490 | |||
| XM_017026566.1 | XP_016882055.1 | 12 | 1423 | |||
| GenBank mRNA | AK128088.1 | - | 16 | 1553 | ||
| BC031233.1 | - | 5 | 495 | |||
| BC043005.1 | - | 16 | 1531 | 5297674 | ||

CATSPERD
Genomic Map