siRNA Details
| Gene Symbol: TMEM130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134450.1 | NP_001127922.1 | 1, 2 | 282 | ||
| NM_152913.2 | NP_690877.1 | 1, 2 | 282 | |||
| GenBank mRNA | AK124634.1 | - | 1, 2 | 1159 | ||
| AK225783.1 | - | 1, 2 | 225 | |||
| AK312364.1 | - | 1, 2 | 152 | |||
| AL834348.1 | - | 1, 2 | 128 | |||
| AL834349.1 | - | 1, 2 | 98 | |||
| AY358583.1 | - | 2, 3 | 214 | |||
| BC030793.1 | - | 1, 2 | 251 | 5263130 | ||
| DA144530.1 | - | 1, 2 | 282 | |||

TMEM130
Genomic Map