siRNA Details
| Gene Symbol: C15orf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301106.1 | NP_001288035.1 | 1 | 814 | ||
| NM_153040.2 | NP_694585.1 | 1 | 814 | |||
| XM_011521257.2 | XP_011519559.1 | 1 | 6729 | |||
| GenBank mRNA | AK057393.1 | - | 1 | 815 | ||
| BC109252.2 | - | 1 | 570 | 40023840 | ||
| BC109253.1 | - | 1 | 568 | 40023844 | ||

C15orf32
Genomic Map