siRNA Details
| Gene Symbol: IQCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305122.1 | NP_001292051.1 | 3 | 921 | ||
| NM_153208.2 | NP_694940.1 | 3 | 921 | |||
| NR_130967.1 | 3 | 921 | ||||
| NR_130968.1 | 3 | 921 | ||||
| XM_011545736.2 | XP_011544038.1 | 2 | 256 | |||
| XM_017022933.1 | XP_016878422.1 | 2 | 967 | |||
| GenBank mRNA | AF520569.1 | - | 3 | 921 | ||
| AF520570.1 | - | 3 | 921 | |||
| AK301933.1 | - | 2 | 244 | |||
| AK311216.1 | - | 2 | 214 | |||
| AK315672.1 | - | 2 | 244 | |||
| BC034823.1 | - | 2 | 252 | 5165639 | ||

IQCK
Genomic Map