siRNA Details
| Gene Symbol: NKAIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152864.3 | NP_690603.3 | 6, 7 | 707 | ||
| XM_017027638.1 | XP_016883127.1 | 6, 7 | 1127 | |||
| XM_017027639.1 | XP_016883128.1 | 6, 7 | 1113 | |||
| GenBank mRNA | AK126246.1 | - | 6, 7 | 630 | ||
| AK225765.1 | - | 6, 7 | 661 | |||
| BC041812.1 | - | 6, 7 | 705 | 5302801 | ||
| BG708360.1 | - | 6, 7 | 710 | |||

NKAIN4
Genomic Map