siRNA Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001719.2 | NP_001710.1 | 2 | 969 | ||
| GenBank mRNA | AK291186.1 | - | 2 | 545 | ||
| AK312419.1 | - | 2 | 542 | |||
| BC008584.1 | - | 2 | 481 | 4183402 | ||
| EU679366.1 | - | 2 | 353 | |||
| M60316.1 | - | 2 | 536 | |||
| X51801.1 | - | 2 | 562 | |||

BMP7
Genomic Map