siRNA Details
| Gene Symbol: KBTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152903.4 | NP_690867.3 | 1 | 1866 | ||
| GenBank mRNA | AK056633.1 | - | 1 | 1811 | ||
| AK095195.1 | - | 1 | 511 | |||
| AK096608.1 | - | 2 | 1806 | |||
| AK299627.1 | - | 2 | 1614 | |||
| AL833839.1 | - | 1 | 1518 | |||
| BC000560.2 | - | 1 | 1788 | 3163484 | ||
| BC051349.1 | - | 1 | 1627 | 6483483 | ||

KBTBD6
Genomic Map