siRNA Details
Gene Symbol: TMEM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153015.2 | NP_694560.1 | 2 | 452 | ||
GenBank mRNA | AK055230.1 | - | 2 | 378 | ||
BC030710.2 | - | 2 | 452 | 4819093 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153015.2 | NP_694560.1 | 2 | 452 | ||
GenBank mRNA | AK055230.1 | - | 2 | 378 | ||
BC030710.2 | - | 2 | 452 | 4819093 |